2-Chloroisonicotinonitrile
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2-Chloro-4-cyanopyridine, 2-Chloropyridine-4-carbonitrile
SPB06434.1 SPB06434.50
MAYBSPB06434.1EA 0 EUR
MAYBSPB06434.1 MAYBSPB06434.50
2-Chloroisonicotinonitrile
2-Chloroisonicotinonitrile
Formule: C₆H₃ClN₂
MW: 138.56 g/mol
MDL Nummer: MFCD00174318
CAS nummer: 33252-30-1
UN: 3439
ADR: 6.1 III