1-(2-Methoxyphenyl)-2-thiourea
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
SPB06629.1
SPB06629.10
SPB06629.50
MAYBSPB06629.1EA
0
EUR
MAYBSPB06629.1
MAYBSPB06629.10
MAYBSPB06629.50
1-(2-Methoxyphenyl)-2-thiourea
1-(2-Methoxyphenyl)-2-thiourea
Formule:
(C₆H₄(OCH₃))NC(S)NH₂ Kookpunt: 310 °C (1013 hPa) Smeltpunt: 156 °C |
MDL Nummer:
MFCD00022162 CAS nummer: 1516-37-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...