α,α,α-Trifluoro-o-toluonitrile

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Cyanobenzotrifluoride, o-(Trifluoromethyl)benzonitrile, 2-(Trifluoromethyl)benzonitrile

TL00228.1 TL00228.10 TL00228.50
MAYBTL00228.1EA 0 EUR
MAYBTL00228.1 MAYBTL00228.10 MAYBTL00228.50
α,α,α-Trifluoro-o-toluonitrile
α,α,α-Trifluoro-o-toluonitrile
Formule: C₈H₄F₃N
Kookpunt: 204 °C (1013 hPa)
Smeltpunt: 18 °C
Dichtheid: 1,2886 g/cm³ (20 °C)
MDL Nummer: MFCD00001791
CAS nummer: 447-60-9
EINECS: 207-184-3
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR