1-Chloroethyl chloroformate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1-Chloroethyl carbonochloridate

C1395-25G C1395-250G
TCIAC1395-25GEA 33 EUR
TCIAC1395-25G TCIAC1395-250G
1-Chloroethyl chloroformate ≥98.0% (by GC)
1-Chloroethyl chloroformate
Formule: C₃H₄Cl₂O₂
MW: 142.97 g/mol
Kookpunt: 120°C
Smeltpunt: -65
Vlampunt: 41°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000647
CAS nummer: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d04df2db900031e94092162eb801ab5f36cb490&vid=8d04df2db900031e94092162eb801ab5f36cb490&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR