Ethylbroomacetaat, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Bromoacetic ethyl ester
M-Clarity™ quality level = MQ200
8.01636.0250 8.01636.1000
8.01636.0250EA 87.1 EUR
8.01636.0250 8.01636.1000
Ethylbroomacetaat, Sigma-Aldrich®
Ethylbroomacetaat
Formule: BrCH₂COOCH₂CH₃
Kookpunt: 159 °C (1013 hPa)
Smeltpunt: –38 °C
Dichtheid: 1,5 g/cm³ (20 °C)
Vlampunt: 58 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000191
CAS nummer: 105-36-2
EINECS: 203-290-9
UN: 1603
ADR: 6.1 (3),II
Order Now
Specification Test Results
Solubility (20 °C) decomposes
Melting Point -38 °C
Molar Mass 167.00 g/mol
Boiling Point 158 - 161 °C (1013 hPa)
Vapor Pressure 4.5 hPa (20 °C)
Flash Point 50 °C
Refractive Index 1.451 (20 °C, 589 nm)
Density 1.50 g/cm3 (20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials