Ethyl crotonate for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.02668.0250 8.02668.1000
8.02668.0250EA 80.4 EUR
8.02668.0250 8.02668.1000
Ethyl crotonate for synthesis, Sigma-Aldrich®
Ethyl crotonate
Formule: C₆H₁₀O₂
Kookpunt: 143 °C (1013 hPa)
Smeltpunt: –79 °C
Dichtheid: 0,9208 g/cm³ (20 °C)
Vlampunt: 2 °C
MDL Nummer: MFCD00009289
CAS nummer: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 13,02625
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Molar Mass 114.14 g/mol
Boiling Point 138 - 140 °C (1013 hPa)
Vapor Pressure 65 hPa (50 °C)
Flash Point 2 °C
Refractive Index 1.4248 (20 °C, 589 nm)
Density 0.92 g/cm3 (20 °C)