1,3-Dimethylurea, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.03244.0100 8.03244.1000
8.03244.0100EA 57 EUR
8.03244.0100 8.03244.1000
1,3-Dimethylurea, Sigma-Aldrich®
1,3-Dimethylurea
Formule: C₃H₈N₂O
Kookpunt: 268 °C (1013 hPa)
Smeltpunt: 96 °C
Dichtheid: 1,142 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008286
CAS nummer: 96-31-1
EINECS: 202-498-7
Order Now
Specification Test Results
Solubility 765 g/l (21.5 °C)
Melting Point 105 - 108 °C
Molar Mass 88.11 g/mol
Bulk Density 500 kg/m3
Boiling Point 268 - 270 °C (1013 hPa)
Vapor Pressure 6 hPa (115 °C)
Density 1.14 g/cm3 (20 °C)
pH 9.0 - 9.5 (100 g/l, H2O, 20 °C)
Ignition Point 400 °C