Furfurylalcohol, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Furfurol
M-Clarity™ quality level = MQ200
8.04015.0100 8.04015.1000
8.04015.0100EA 34.2 EUR
8.04015.0100 8.04015.1000
Furfurylalcohol, Sigma-Aldrich®
Furfurylalcohol
Formule: C₅H₆O₂
MW: 98.1 g/mol
Kookpunt: 171
Smeltpunt: –31 °C
Dichtheid: 1.13 g/cm³ (20 °C)
Vlampunt: 75 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003252
CAS nummer: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1 III
Merck Index: 13,04326
Order Now
Specification Test Results
Hill Formula C₅H₆O₂
Molar Mass 98.1 g/mol
Boiling point 170 - 171 °C (1013 mbar)
Density 1.13 g/cm³ (20 °C)
Explosion limit 1.8 - 16.3 %(V)
Flash point 65 °C
Ignition temperature 390 °C
Melting Point -31 °C
pH value 6 (300 g/l, H₂O, 20 °C)
Vapor pressure 0.53 hPa (20 °C)
Refractive Index 1.4845 (20 °C, 589 nm)
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 177 mg/kg
LD 50 dermal LD50 Rabbit 400 mg/kg