Tetrabutylammonium hydroxide 20% in aqueous solution for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.18759.0100 8.18759.0500 8.18759.0050
8.18759.0100EA 67.9 EUR
8.18759.0100 8.18759.0500 8.18759.0050
Tetrabutylammonium hydroxide 20% in aqueous solution for synthesis, Sigma-Aldrich®
Tetrabutylammonium hydroxide
Formule: C₁₆H₃₇NO
MW: 259.48 g/mol
Smeltpunt: 29.85
Dichtheid: 0.968
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Boiling Point 102 °C (1013 hPa)
Density 0.99 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)