Water Reag. Ph. Eur. 1095506 nitrate-free
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87961.290EA
35.4
EUR
87961.290
Water Reag. Ph. Eur. 1095506 nitrate-free
Water
Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formule:
H₂O MW: 18,02 g/mol Kookpunt: 100 °C (1013 hPa) Smeltpunt: 0 °C Dichtheid: 1 g/cm³ (4 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011332 CAS nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...