N-Ethylmaleimide (NEM)
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione
11331.02
11331.03
SERA11331.02EA
0
EUR
SERA11331.02
SERA11331.03
N-Ethylmaleimide (NEM)
N-Ethylmaleimide (NEM)
Formule:
C₆H₇NO₂ MW: 125.13 g/mol Kookpunt: 210 Smeltpunt: 46 |
MDL Nummer:
MFCD00005509 CAS nummer: 128-53-0 EINECS: 204-892-4 UN: 2928 ADR: 6.1 II Merck Index: 13,03855 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...