Benzidine

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 4,4'-Diaminobiphenyl, 4-(4-Aminophenyl)aniline, 1,1’-Biphenyl-4,4’-diamine, Biphenyl-4,4'-ylenediamine

14542.01 14542.02
SERA14542.01EA 0 EUR
SERA14542.01 SERA14542.02
Benzidine
Benzidine
Formule: C₁₂H₁₂N₂
MW: 184.24 g/mol
Kookpunt: 359
Smeltpunt: 126…127 °C
Dichtheid: 1.251 g/cm³ (20 °C)
MDL Nummer: MFCD00039765
CAS nummer: 92-87-5
EINECS: 202-199-1
UN: 1885
ADR: 6.1 II
Merck Index: 12,01106

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR