Tricine

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

37195.01 37195.03
SERA37195.01EA 0 EUR
SERA37195.01 SERA37195.03
Tricine
Tricine
Formule: (HOCH₂)₃CNHCH₂CO₂H
Smeltpunt: 187 °C
Dichtheid: 1,523 g/cm³ (19.85 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004277
CAS nummer: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR