2,2,2-Trichloroethyl chloroformate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C0795-250G C0795-25G
TCIAC0795-250GEA 757 EUR
TCIAC0795-250G TCIAC0795-25G
2,2,2-Trichloroethyl chloroformate ≥98.0% (by GC)
2,2,2-Trichloroethyl chloroformate
Formule: C₃H₂Cl₄O₂
Kookpunt: 172°C
Smeltpunt: 1 °C
Dichtheid: 1,53 g/cm³ (20 °C)
MDL Nummer: MFCD00000810
CAS nummer: 17341-93-4
EINECS: 241-363-7
UN: 3277
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2d10fa30225cd48ed02f6a524eabb6c9e8e3c1&vid=ee2d10fa30225cd48ed02f6a524eabb6c9e8e3c1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %