N,N,N-Trimethylanilinium tribromide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

P0928-25G P0928-500G
TCIAP0928-25GEA 42.9 EUR
TCIAP0928-25G TCIAP0928-500G
N,N,N-Trimethylanilinium tribromide ≥98.0% (by titrimetric analysis)
N,N,N-Trimethylanilinium tribromide
Formule: C₉H₁₄Br₃N
MW: 375.93 g/mol
MDL Nummer: MFCD00011789
CAS nummer: 4207-56-1
EINECS: 224-127-8
UN: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2659e61e7f3f02cbb474d3fdf3c29e7a706f3d9&vid=f2659e61e7f3f02cbb474d3fdf3c29e7a706f3d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Melting point 113.0 to 118.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR