Bacteria prepared slides
Leverancier: LIEDER JOHANNES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
191080-100 191080-101 191080-102 191080-103 BA131D
763-0361EA 142 EUR
763-0361 763-0362 763-0363 763-0364 763-0366
Bacteria prepared slides
Object- en dekglaasjes Voorbereide dia's
This bacteriological slide includes.
  • Streptococcus
  • Diphtheria bacilli
  • Spirillum
  • Staphylococcus
  • Oral flora
  • Septic arthritis
  • Syphilis
  • Tuberculosis (Tb)
  • Tetanus germs
  • Coliform
  • Salmonella germs
  • Typhoid fever germs
Leveringsinformatie: Supplied with pictures and explanatory texts.