Broad range hydrometers
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3000TB100/20QP 3050TB065/20QP 3050FG100/20QP
765-0052EA 16.7 EUR
765-0052 765-0053 ALLA3050FG100/20QP
Broad range hydrometers
Hydrometers
These broad range hydrometers are calibrated at 20 °C.
Mostly dedicated to applications with citric and tartaric acids.