Anti-ATM (Ser1981) phosphate Mouse Monoclonal Antibody [clone: '10H11.E12]

Leverancier: Cell Signaling
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4526L 4526S
CSTA4526LEA 1161 EUR
CSTA4526L CSTA4526S
Anti-ATM (Ser1981) phosphate Mouse Monoclonal Antibody [clone: '10H11.E12]
Antilichamen
Type: Primary Antibodies
Antigen: ATM (Ser1981) phosphate
Clonality: Monoclonal
Clone: '10H11.E12
Conjugation:
Epitope:
Hoat: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR