Anti-Cdc2 (Tyr15) phosphate Rabbit Monoclonal Antibody [clone: 10A11]

Leverancier: Cell Signaling
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4539L 4539S
CSTA4539LEA 1219 EUR
CSTA4539L CSTA4539S
Anti-Cdc2 (Tyr15) phosphate Rabbit Monoclonal Antibody [clone: 10A11]
Antilichamen
Type: Primary
Antigen: Cdc2 (Tyr15) phosphate
Clonality: Monoclonal
Clone: 10A11
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR