Anti-Lysine (K-Ac) acetylate Rabbit Polyclonal Antibody

Leverancier: Cell Signaling
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

9441L 9441S
CSTA9441LEA 1211 EUR
CSTA9441L CSTA9441S
Anti-Lysine (K-Ac) acetylate Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Lysine (K-Ac) acetylate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Any species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR