Anti-H3 (Lys9/Lys14) Acetylate Rabbit Polyclonal Antibody

Leverancier: Cell Signaling
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

9677L 9677S
CSTA9677LEA 1033 EUR
CSTA9677L CSTA9677S
Anti-H3 (Lys9/Lys14) Acetylate Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: H3 (Lys9/Lys14) acetylate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Monkey, S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR