HistoBond® Adhesive Microscope Slides with Circles
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0810471 0810461
MARI0810471EA 49.9 EUR
MARI0810471 MARI0810461
HistoBond® Adhesive Microscope Slides with Circles
Object- en dekglaasjes Adhesie-objectglaasjes
For use in cytocentrifuges we offer our adhesive and positively charged HistoBond®+ slides with white rings. These serve as a microscope help to find the centrifuged cells more easily.
  • Made of soda lime glass of third hydrolytic class
  • Dimensions: ~76×26 mm
  • Thickness: ~ 1 mm
  • With white marking area of ~20 mm on one side
  • Pre-cleaned and ready for use
  • Autoclavable
For applications according to EU regulation 2017/746 on invitro diagnostic medical devices (IVDR), with CE-mark, recommended best before date and batch number for comprehensive information and traceability. Suitable for marking with various printer systems and can be written with permanent markers (like laboratory marker Cat. No. 6130603).
Certificaten: DIN ISO8037-1 certified.
Verpakking: In plastic boxes of 100 pieces, 20 boxes in a carton.