Scepter™ M321 Sterile Glove Liners
Leverancier: WELLS LAMONT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M321EU-S M321EU-M M321EU-L M321EU-XL
112-6621EA 341 EUR
112-6621 112-6620 112-6619 112-6622
Scepter™ M321 Sterile Glove Liners
Handschoenen
Sterile and reusable cut-resistant glove liners.
  • For use in cleanroom environments
  • Antimicrobial
Scepter™ cut-resistant liners protect hands from prolonged exposure to latex and offer a high level of tactile sensation. The M321 is lightweight and sterile in addition to providing cut and abrasion resistance due to its EN 388 1X31D rating. Scepter™ gloves can be used as a liner or stand-alone.
Certificaten: EN 388 1X31D. Category III for particulates.
Verpakking: Gloves are packaged in a box of 10 pairs. Each pair is sealed in its own sterile pouch.