VWR® Anti-Vibration Tables
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
611-5596EA 1400 EUR
611-5596 611-5597
VWR® Anti-Vibration Tables
Meubilair Tafels
The anti-vibration table provides a stable and vibration-free surface for sensitive equipment, such as microscopes, balances, or other precision instruments. It helps to minimise external vibrations from affecting the performance and accuracy of the equipment, ensuring reliable measurements and observations.
  • The integrated granite isolation area sits flush with the surrounding work area (standard), maximising available space
  • Provides suitable working conditions for sensitive devices
  • Large working surface of 900×600 mm with integrated granite slab (500×350 mm) ideal for balances and microscopes
  • High resistant phenolic laminate worktop and powder-coated steel frame are durable and easy to clean
The anti-vibration table standard is made by two separate metal steel frame: outer structure to isolate the working area and the inner structure that holds the granite slab. The granite slab is held by four high performance special dampers. The structure is made of powder-coated steel supported by vibration absortpion feet and the table top is made by high resistant phenolic laminate.
The anti-vibration table advanced is made of a solid large granite slab leaned on two solid granite legs with an outer separate metal steel frame. The granite top is held by four ball-head solid stainless steel support. The outer structure is made of powder-coated steel. The table top is made by high resistant phenolic laminate.
Bestelinformatie: Installation not included.