Molecular model: electron repulsion
Leverancier: Spiring Enterprises
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
761-0116EA 0 EUR
761-0116
Molecular model: electron repulsion
Modellen Moleculaire biologie modellen
Lone pairs of electrons are represented with brown balls.
Two additional pear-shaped electron clouds are included to demonstrate the hydronium ion that forms from a hydrogen ion from hydrochloric acid.