ESD Safety Shoes, Low, ENNO XXT Pro, S3S
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720311/46 720311/47 720311/39 720311/40 720311/41 720311/42 720311/43 720311/44 720311/45
ELTN720311/46EA 96.9 EUR
ELTN720311/46 ELTN720311/47 ELTN720311/39 ELTN720311/40 ELTN720311/41 ELTN720311/42 ELTN720311/43 ELTN720311/44 ELTN720311/45
ESD Safety Shoes, Low, ENNO XXT Pro, S3S
Schoenen en laarzen
Hydrophobised textile material with recycled material content/hydrophobised microfibre.
  • Breathable lining with recycled material content
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black with recycled material content
  • Non-metallic, penetration-resistant midsolewith recycled material content
  • PU/PU sole WELLMAXX TRAINERS
  • TPU scuff cap
  • Composite toe cap cwith recycled material content
Designed for use in areas where there is a risk of electrostatic discharge (ESDS/ESD) and workplaces on hard undergrounds. The revolutionary Infinergy® soleMore Product Information