(R)-(-)-Linalool 95% (sum of enantiomers)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool
428311000 428310250
ACRO428311000EA 90.7 EUR
ACRO428311000 ACRO428310250
(R)-(-)-Linalool 95% (sum of enantiomers)
(R)-(-)-Linalool
Formule: C₁₀H₁₈O
MW: 154.25 g/mol
Kookpunt: 196…198 °C (760 mmHg)
Dichtheid: 0.862 g/cm³
Vlampunt: 75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00135469
CAS nummer: 126-91-0
EINECS: 204-811-2
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4640 (20°C, 589 nm)
GC ≥94 %
Specific gravity 0.859 to 0.864
Specific optical rotation -15° to -21° (20°C, 589 nm) (neat)