Triethylene glycol 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol
139590010 139590025 139590100
ACRO139590010EA 58.4 EUR
ACRO139590010 ACRO139590025 ACRO139590100
Triethylene glycol 99%
Triethylene glycol
Formule: C₆H₁₄O₄
MW: 150.17 g/mol
Kookpunt: 285 °C (760 mmHg)
Smeltpunt: –7 °C
Dichtheid: 1.12 g/cm³
Vlampunt: 165 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002880
CAS nummer: 112-27-6
EINECS: 203-953-2
Merck Index: 12,09802
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4545 to 1.4565 (20°C, 589 nm)
GC ≥98.5 %