Triethylene glycol

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol

4520.1 4520.2 4520.3
ROTH4520.1EA 0 EUR
ROTH4520.1 ROTH4520.2 ROTH4520.3
Triethylene glycol
Triethylene glycol
Formule: C₆H₁₄O₄
MW: 150.17 g/mol
Kookpunt: 285
Smeltpunt: –7 °C
Dichtheid: 1.125 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002880
CAS nummer: 112-27-6
EINECS: 203-953-2
Merck Index: 12,09802

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR