Triethylene glycol, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.08245.1000 8.08245.9220
8.08245.1000EA 46 EUR
8.08245.1000 8.08245.9220
Triethylene glycol, Sigma-Aldrich®
Triethylene glycol
Formule: C₆H₁₄O₄
Kookpunt: 285 °C (1013 hPa)
Smeltpunt: –7 °C
Dichtheid: 1,125 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002880
CAS nummer: 112-27-6
EINECS: 203-953-2
Merck Index: 12,09802
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -7 °C
Molar Mass 150.17 g/mol
Boiling Point 285 - 295 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 165 °C
Refractive Index 1.4559 (20 °C, 589 nm)
Explosion Limit 0.9 - 9.2 %(V)
Density 1.123 g/cm3 (20 °C)
pH 6.5 - 7.5 (100 g/l, H2O, 20 °C)
Ignition Point 370 °C