N,N-Diethylethylenediamine 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-(Diethylamino)ethylamine, 2-Aminoethyldiethylamine
A19636.22 A19636.36
A19636.22EA 35.9 EUR
A19636.22 A19636.36
N,N-Diethylethylenediamine 99%
N,N-Diethylethylenediamine
Purity: 99% CAS No.: 100-36-7 Molecular Formula: (CH3CH2)2NCH2CH2NH2 Formula Weight: 116.21 Flash Point: 30°(86°F) Refractive Index: 1.4360 Melting Point: -70° Boiling Point: 145-147° Beilstein Reference: 605299 UN No.: 2685 MDL No.: MFCD00008176
Formule: C₆H₁₆N₂
MW: 116.21 g/mol
Kookpunt: 145…147 °C
Smeltpunt: < –70 °C
Dichtheid: 0.825
Vlampunt: 30 °C (86 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008176
CAS nummer: 100-36-7
EINECS: 202-844-7
UN: 2685
ADR: 8 II