N,N-Diethyl-1,3-diaminopropaan, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 3-Diethylamino-1-propylamine, DEAPA, N,N-Diethyl-1,3-propanediamine, N,N-Diethyl-1,3-diaminopropane, 3-Aminopropyldiethylamine, 3-(Diethylamino)propylamine
M-Clarity™ quality level = MQ200
8.14100.0100 8.14100.1000 8.14100.9020
8.14100.0100EA 35.1 EUR
8.14100.0100 8.14100.1000 8.14100.9020
N,N-Diethyl-1,3-diaminopropaan, Sigma-Aldrich®
N,N-Diethyl-1,3-diaminopropaan
Formule: C₇H₁₈N₂
Kookpunt: 168…171 °C (1013 hPa)
Smeltpunt: –60 °C
Dichtheid: 0,8283 g/cm³ (20 °C)
Vlampunt: 51 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008218
CAS nummer: 104-78-9
EINECS: 203-236-4
UN: 2684
ADR: 3 (8),III
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -60 °C
Molar Mass 130.23 g/mol
Boiling Point 159.0 °C
Vapor Pressure 2.2 hPa (20 °C)
Flash Point 51 °C
Refractive Index 1.4413 (20 °C, 589 nm)
Explosion Limit 1.0 %(V)
Density 0.82 g/cm3 (20 °C)
pH 11.7 (10 g/l, H2O, 20 °C)
Ignition Point 215 °C