4-Chloor-3,5-dimethylfenol 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol
109061000 109065000 109060025
ACRO109061000EA 20 EUR
ACRO109061000 ACRO109065000 ACRO109060025
4-Chloor-3,5-dimethylfenol 99%
4-Chloor-3,5-dimethylfenol
Formule: C₈H₉ClO
MW: 156.61 g/mol
Kookpunt: 246 °C (760 mmHg)
Smeltpunt: 113…116 °C
Vlampunt: 132 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002324
CAS nummer: 88-04-0
EINECS: 201-793-8
UN: 3077
ADR: 9 III
Merck Index: 13,02194
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 113 to 116 °C
GC ≥98.5 %
Stabilizer ca. 500 ppm MEHQ