Nitrilotriacetic acid 99
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: NTA, 2,2',2''-Nitrilotriacetic acid
A11936.0E A11936.22 A11936.36
A11936.0EEA 380 EUR
A11936.0E A11936.22 A11936.36
Nitrilotriacetic acid 99
Nitrilotriacetic acid
Purity: 99% CAS No.: 139-13-9 Molecular Formula: C6H9NO6 Formula Weight: 191.14 Odour: Odourless Melting Point: 246° dec. Beilstein Reference: 1710776 Merck Reference: 14,6579 MDL No.: MFCD00004287
Formule: C₆H₉NO₆
MW: 191.14 g/mol
Kookpunt: 498
Smeltpunt: 246 °C (ontbinding)
Dichtheid: 1.67 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004287
CAS nummer: 139-13-9
EINECS: 205-355-7
UN: 2923
ADR: 8 II
Merck Index: 12,06675