2,2'-Thiodiacetic acid, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Thiodiglycolic acid, Thiodiacetic Acid
M-Clarity™ quality level = MQ200
8.08625.0005 8.08625.0250 8.08625.1000
8.08625.0005EA 0 EUR
8.08625.0005 8.08625.0250 8.08625.1000
2,2'-Thiodiacetic acid, Sigma-Aldrich®
2,2'-Thiodiacetic acid
Formule: C₄H₆O₄S
Kookpunt: 168 °C (14 mmHg)
Smeltpunt: 128…131 °C
Dichtheid: 1,661 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004359
CAS nummer: 123-93-3
EINECS: 204-663-9
UN: 3261
ADR: 8,II
Merck Index: 12,09467
Order Now
Specification Test Results
Melting Point 128-131°C
Hazard Class 8
Hazard Class Description Corrosive Material