2,4-Dibromo-3-pentanone mixture of stereoisomers ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A16680.09
A16680.18
A16680.09EA
101
EUR
A16680.09
A16680.18
2,4-Dibromo-3-pentanone mixture of stereoisomers ≥97%
2,4-Dibromo-3-pentanone
Purity: 97%
CAS No.: 815-60-1
Molecular Formula: C5H8Br2O
Formula Weight: 243.91
Storage Temperature: 2°C to 8°C
Odour: Pungent
Boiling Point: 67-82°/10mm
UN No.: 3265
MDL No.: MFCD00015719
Formule:
H₃CBrCHC(O)BrCHCH₃ Kookpunt: 67…82 °C (10 mmHg) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00015719 CAS nummer: 815-60-1 UN: 3265 ADR: 8,II |