4-(Bromomethyl)phenylboronic acid tech. 85%, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L19953.03
L19953.06
L19953.03EA
89
EUR
L19953.03
L19953.06
4-(Bromomethyl)phenylboronic acid tech. 85%, Technical Grade
4-(Bromomethyl)phenylboronic acid
Purity: 85%
CAS No.: 68162-47-0
Molecular Formula: C7H8BBrO2
Formula Weight: 214.85
Melting Point: ca 145-150°
Beilstein Reference: 3030979
UN No.: 3261
MDL No.: MFCD01318113
Formule:
C₇H₈BBrO₂ Smeltpunt: ∼ 145…150 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD01318113 CAS nummer: 68162-47-0 UN: 3261 ADR: 8,III |