4-Iodophenylboronic acid 97%
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (4-Iodophenyl)boronic acid, Dihydroxy(4-iodophenyl)borane, 4-Iodobenzeneboronic acid, (4-Iodophenyl)metaboric acid
AC35948.10 359480010 359480050 359480250
MAYBAC35948.10EA 0 EUR
MAYBAC35948.10 ACRO359480010 ACRO359480050 ACRO359480250
4-Iodophenylboronic acid 97%
4-Iodophenylboronic acid
Formule: C₆H₆BIO₂
MW: 247.83 g/mol
Smeltpunt: 322…326 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01319014
CAS nummer: 5122-99-6
Order Now
Specification Test Results
Appearance Light beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %