6-Methylnicotinic acid ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

L08877.03 L08877.06 L08877.14
L08877.03EA 28.6 EUR
L08877.03 L08877.06 L08877.14
6-Methylnicotinic acid ≥99%
6-Methylnicotinic acid

Purity: 99% CAS No.: 3222-47-7 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: 210-215° Beilstein Reference: 112049

Formule: C₇H₇NO₂
Smeltpunt: 210…215 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006341
CAS nummer: 3222-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR