(±)-Pipecolinic acid
Leverancier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4030766.0025
4030766.0100
BACE4030766.0025EA
0
EUR
BACE4030766.0025
BACE4030766.0100
(±)-Pipecolinic acid
(±)-Pipecolinic acid
Formule:
C₆H₁₁NO₂ |
MDL Nummer:
MFCD00064347 CAS nummer: 535-75-1 EINECS: 208-616-3 |