(±)-Nipecotic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

AC19919DA AC19919EA AC19919EE
MAYBAC19919DAEA 0 EUR
MAYBAC19919DA MAYBAC19919EA MAYBAC19919EE
(±)-Nipecotic acid
(±)-Nipecotic acid
Formule: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Nummer: MFCD00005992
CAS nummer: 498-95-3
EINECS: 207-873-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR