D(+)-Sorbose ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B21208.02
B21208.77
B21208.02EA
368
EUR
B21208.02
B21208.77
D(+)-Sorbose ≥98%
D-(+)-Sorbose
Purity: 98%
CAS No.: 3615-56-3
Molecular Formula: C6H12O6
Formula Weight: 180.16
Melting Point: 163-165°
MDL No.: MFCD00151095
Formule:
C₆H₁₂O₆ Kookpunt: 380 °C (1013 hPa) Smeltpunt: 163…165 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00151095 CAS nummer: 3615-56-3 EINECS: 222-796-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...