Dithiodiglycol (bis(2-hydroxyethyl) disulphide) tech. 90%, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-Hydroxyethyl disulphide, 2,2'-Dithiobisethanol, 2,2'-Dithiodiethanol, Bis(2-hydroxyethyl) disulphide
L11800.18 L11800.30
L11800.18EA 64.6 EUR
L11800.18 L11800.30
Dithiodiglycol (bis(2-hydroxyethyl) disulphide) tech. 90%, Technical Grade
Dithiodiglycol (bis(2-hydroxyethyl) disulphide)
Purity: 90% CAS No.: 1892-29-1 Molecular Formula: C4H10O2S2 Formula Weight: 154.24 Flash Point: 110°(230°F) Refractive Index: 1.5700 Odour: Unpleasant Melting Point: ca 25° Boiling Point: 158-163°/3.5mm Beilstein Reference: 1735851 UN No.: 2810 MDL No.: MFCD00002906
Formule: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Kookpunt: 158…163 °C (3.5 mmHg)
Smeltpunt: ∼ 25 °C
Dichtheid: 1.261
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002906
CAS nummer: 1892-29-1
EINECS: 217-576-6
UN: 2810
ADR: 6.1 III