Combitips® advanced Biopur® Individually Blister-Wrapped Positive Displacement Pipette Tips
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0030089.634 0030089.642 0030089.626 0030089.618 0030089.669 0030089.677 0030089.650 0030089.693 0030089.685
613-2069EA 202 EUR
613-2069 613-2070 613-2068 613-2067 613-2072 613-2073 613-2071 613-2075 613-2074
Combitips® advanced Biopur® Individually Blister-Wrapped Positive Displacement Pipette Tips
Pipettips
Eppendorf Combitips® advanced, individually blister-wrapped in 'Biopur®' grade, offer unparalleled purity for critical lab tasks, ensuring contamination-free, precise pipetting with an ergonomic design. Ideal for highly sensitive molecular biology applications.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • Nine volume sizes (0,1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters
  • Combitips® advanced in 'Biopur' quality are guaranteed to be sterile and free of pyrogens, RNases, DNases, DNA, ATP, and PCR inhibitors
  • Lot-specific certificates for Eppendorf consumables in 'Biopur®' quality are available
  • Elongated Combitips® advanced in volumes 2,5, 5,0, and 10 ml will reach the bottom of most common laboratory vessels
  • Combitips® advanced are offered in five different purity grades to support a range of applications
  • Appropriate purity grade for each application: Eppendorf Quality, PCR clean, Forensic DNA Grade, sterile, or Biopur®
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
"The tip of choice when working with Repeater®/Multipette® multi-dispenser pipettes from Eppendorf:
Thanks to their revolutionary design, Combitips® advanced dispenser tips provide many benefits for work in a modern laboratory. With an integrated piston, they function according to the positive displacement principle and ensure that the correct volume of liquid is dispensed regardless of the physical properties of the liquid (e.g., viscosity, volatility, density, temperature). Achieve high-precision dispensing, independent of the flow characteristics of the liquid, and safely work with radioactive or toxic materials due to the hermetically sealed piston that prevents aerosol contamination.
Eppendorf pioneered the Repeater®/Combitips® multi-dispenser system in 1978 and improved it several times since. It was the very first pipetting system allowing users to fill the tip once and dispense multiple times. The concept remains a driver for efficient liquid handling and a staple in many labs working with plates or series of tubes. In 2023, the Combitips® advanced pipette tip line-up has been certified with ACT® Environmental Impact Factor Labels by the My Green Lab® non-profit organization, making it the ideal choice for labs seeking more sustainable products.
Certificaten: ACT Label and Lot-specific certficates available for the purity grade Biopur can be downloaded from the Eppendorf website.
Bestelinformatie: Please note: Combitips® advanced are designed for use with Eppendorf Repeater®/Multipette® multi-dispensers only.