Rotors for Vacufuge Plus
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5490030.001 5490032.004 5490034.007 5490035.003 5490036.000 5490037.006 5490038.002 5490039.009 5490040.007 5490045.009 5490042.000 5490041.003 5490043.006 5490047.001 5490048.008
700-1381EA 449 EUR
700-1381 700-1383 700-1382 522-1833 522-1834 521-0119 522-1836 522-1837 522-1838 522-1895 521-0121 521-0120 521-0122 521-2070 EPPE5490048.008
Rotors for Vacufuge Plus
Centrifuge Rotoren
Eppendorf Vacufuge Plus efficiency meets precision. Tailor your concentration processes with a max speed of 1700 rpm. Accommodate diverse samples with a 48×1,5/2,0 ml capacity. Customise your setup with various accessories for optimal sample concentration.
  • A wide range of fixed-angle rotors and the microplate rotor are available to accommodate a variety of tubes, plates, and applications
  • Increase capacity with a spacer to stack some fixed-angle rotors
  • Precision engineered for optimal sample concentration
Revolutionise your sample concentration with the Eppendorf Vacufuge Plus. Operating at a max speed of 1700 rpm, this centrifuge concentrator offers versatility for DNA/RNA, protein purification, and more. The 48×1,5/2,0 ml capacity caters to various sample volumes, ensuring efficient concentration. Customise your workflow with accessories like rotors and adaptors. The user-friendly interface and compact design make the Vacufuge plus an essential tool for any lab. Elevate your concentration processes with precision and adaptability.