1,1,1,3,3,3-Hexafluoro-2-propanol, LiChropur™ LC-MS, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP
18127-50ML 18127-10ML
SUPL18127-50MLEA 702 EUR
SUPL18127-50ML 18127-10ML
1,1,1,3,3,3-Hexafluoro-2-propanol, LiChropur™ LC-MS, Supelco®
1,1,1,3,3,3-Hexafluoro-2-propanol
Formule: C₃H₂F₆O
MW: 168.04 g/mol
Kookpunt: 59.1
Smeltpunt: -96
MDL Nummer: MFCD00011651
CAS nummer: 920-66-1
Merck Index: 13,09598