Sephadex® G-100, superfine, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SUPLG10050-10GEA 217 EUR
SUPLG10050-10G
Sephadex® G-100, superfine, Supelco®
Sephadex® G-100
MDL Nummer: MFCD00132235
CAS nummer: 9050-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR