Accessories for Vivapore® Solvent Absorption Concentrators
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VP-ARV0010-D VP-AST0001-C
SARTVP-ARV0010-DEA 30.6 EUR
SARTVP-ARV0010-D SARTVP-AST0001-C
Accessories for Vivapore® Solvent Absorption Concentrators
Concentratoren
Stand with simple push-fit handling, each stand can hold up to four Vivapore units during use.
  • Accessories for use with Vivapore® solvent absorption concentrators
  • Stands hold up to four Vivapore® units during use
  • Expansion reservoirs for increased sample capacity or diafiltration  
Compatible with Vivapore® 5 and 10 solvent absorption concentrators.