SureSTART™ Crimp Caps, 11 mm, Level 2 High-Throughput Applications
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6ACC11NR1 6ACC11NR1B 6ACC11NR1G 6ACC11NR1R 6ACC11NR1Y
HYPE6ACC11NR1EA 18.8 EUR
HYPE6ACC11NR1 HYPE6ACC11NR1B HYPE6ACC11NR1G HYPE6ACC11NR1R HYPE6ACC11NR1Y
SureSTART™ Crimp Caps, 11 mm, Level 2 High-Throughput Applications
Sluitingen Sluitingen voor chromatografievials
These SureSTART™ crimp caps have an 11 mm opening. Select level 2 products for high-throughput applications, when robustness and reproducibility are key.
  • Single-use
  • Not Autoclavable
  • Non sterile
SureSTART™ Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.