Anti-Cathepsin D Rabbit Polyclonal Antibody
Leverancier: United States Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBI309372-100EA 608.01 EUR
USBI309372-100 USBI309369-100
Anti-Cathepsin D Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Cathepsin D
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Hamster